Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VMA22
Entrez Gene ID
856457
Gene Name
Gene Aliases
CEV1, VPH6, YHR060W
Chromosome Location
Chr.VIII: 220725 - 221270 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179190.1 NP_011927.1 1 - 1 452 70

View Full Product Details