Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IPI1
Entrez Gene ID
856485
Gene Name
Gene Aliases
YHR085W
Chromosome Location
Chr.VIII: 276764 - 277768 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179215.1 NP_011953.1 1 - 1 951 92

View Full Product Details