Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDT80
Entrez Gene ID
856524
Gene Name
Gene Aliases
YHR124W
Chromosome Location
Chr.VIII: 356561 - 358444 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179254.1 NP_011992.1 1 - 1 1649 97

View Full Product Details