Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEX18
Entrez Gene ID
856565
Gene Name
Gene Aliases
YHR160C
Chromosome Location
Chr.VIII: 419221 - 420072 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179291.1 NP_012030.1 1 - 1 753 106

View Full Product Details