Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DBP8
Entrez Gene ID
856574
Gene Name
Gene Aliases
YHR169W
Chromosome Location
Chr.VIII: 442181 - 443476 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179300.1 NP_012039.1 1 - 1 1154 88

View Full Product Details