Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRG1
Entrez Gene ID
856616
Gene Name
Gene Aliases
YHR209W
Chromosome Location
Chr.VIII: 519437 - 520312 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179340.1 NP_012079.1 1 - 1 751 90

View Full Product Details