Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YRB2
Entrez Gene ID
854747
Gene Name
Gene Aliases
YIL063C
Chromosome Location
Chr.IX: 242761 - 243744 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179413.1 NP_012201.1 1 - 1 754 98

View Full Product Details