Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLM1
Entrez Gene ID
854701
Gene Name
Gene Aliases
LIT2, YIL105C
Chromosome Location
Chr.IX: 167581 - 169641 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179453.1 NP_012161.1 1 - 1 1656 74

View Full Product Details