Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYR1
Entrez Gene ID
853452
Gene Name
Gene Aliases
CDC35, FIL1, HSR1, SRA4, TSM0185, YJL005W
Chromosome Location
Chr.X: 425157 - 431237 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181439.3 NP_012529.3 1 - 1 5844 80

View Full Product Details