Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LSB6
Entrez Gene ID
853345
Gene Name
Gene Aliases
YJL100W
Chromosome Location
Chr.X: 237263 - 239086 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181533.1 NP_012435.1 1 - 1 1752 94

View Full Product Details