Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
URA2
Entrez Gene ID
853311
Gene Name
Gene Aliases
YJL130C
Chromosome Location
Chr.X: 165723 - 172367 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181563.2 NP_012405.2 1 - 1 6551 103

View Full Product Details