Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MNN11
Entrez Gene ID
853256
Gene Name
Gene Aliases
YJL183W
Chromosome Location
Chr.X: 84068 - 85336 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181616.2 NP_012352.2 1 - 1 1150 75

View Full Product Details