Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GON7
Entrez Gene ID
853255
Gene Name
Gene Aliases
LDB6, PCC2, YJL184W
Chromosome Location
Chr.X: 83448 - 83819 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181617.1 NP_012351.1 1 - 1 247 89

View Full Product Details