Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
JSN1
Entrez Gene ID
853553
Gene Name
Gene Aliases
PUF1, YJR091C
Chromosome Location
Chr.X: 595061 - 598336 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181748.1 NP_012624.1 1 - 1 3149 70

View Full Product Details