Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ILM1
Entrez Gene ID
853582
Gene Name
Gene Aliases
YJR118C
Chromosome Location
Chr.X: 643490 - 644101 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181776.3 NP_012652.3 1 - 1 156 71

View Full Product Details