Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MCM22
Entrez Gene ID
853599
Gene Name
Gene Aliases
YJR135C
Chromosome Location
Chr.X: 676059 - 676778 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181793.1 NP_012669.1 1 - 1 451 83

View Full Product Details