Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IML1
Entrez Gene ID
853603
Gene Name
Gene Aliases
SEA1, YJR138W
Chromosome Location
Chr.X: 684567 - 689321 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181796.2 NP_012672.1 1 - 1 4548 84

View Full Product Details