Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BYE1
Entrez Gene ID
853865
Gene Name
Gene Aliases
YKL005C
Chromosome Location
Chr.XI: 433093 - 434877 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179571.3 NP_012921.3 1 - 1 1444 74

View Full Product Details