Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAB3
Entrez Gene ID
853774
Gene Name
Gene Aliases
YKL088W
Chromosome Location
Chr.XI: 275283 - 276998 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179654.1 NP_012835.1 1 - 1 1446 90

View Full Product Details