Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CWP1
Entrez Gene ID
853766
Gene Name
Gene Aliases
YJU1, YKL096W
Chromosome Location
Chr.XI: 261132 - 261851 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179662.1 NP_012827.1 1 - 1 650 96

View Full Product Details