Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYO3
Entrez Gene ID
853729
Gene Name
Gene Aliases
YKL129C
Chromosome Location
Chr.XI: 196349 - 200167 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179695.2 NP_012793.2 1 - 1 3651 66

View Full Product Details