Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRP49
Entrez Gene ID
853687
Gene Name
Gene Aliases
YKL167C, mL61
Chromosome Location
Chr.XI: 133721 - 134134 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179733.1 NP_012754.1 1 - 1 250 100

View Full Product Details