Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KTR2
Entrez Gene ID
853935
Gene Name
Gene Aliases
YKR061W
Chromosome Location
Chr.XI: 557677 - 558954 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179851.3 NP_012987.3 1 - 1 1151 103

View Full Product Details