Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRP16
Entrez Gene ID
853961
Gene Name
Gene Aliases
PRP23, RNA16, YKR086W
Chromosome Location
Chr.XI: 599857 - 603072 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179876.3 NP_013012.3 1 - 1 2943 94

View Full Product Details