Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BPT1
Entrez Gene ID
850645
Gene Name
Gene Aliases
YLL015W
Chromosome Location
Chr.XII: 116432 - 121111 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181835.1 NP_013086.1 1 - 1 4545 85

View Full Product Details