Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GTT2
Entrez Gene ID
850666
Gene Name
Gene Aliases
YLL060C
Chromosome Location
Chr.XII: 21138 - 21839 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181880.1 NP_013040.1 1 - 1 654 68

View Full Product Details