Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRN2
Entrez Gene ID
850810
Gene Name
Gene Aliases
SRN10, VPL16, VPS37, YLR119W
Chromosome Location
Chr.XII: 385534 - 386175 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182006.1 NP_013220.1 1 - 1 547 89

View Full Product Details