Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STM1
Entrez Gene ID
850843
Gene Name
Gene Aliases
MPT4, YLR150W
Chromosome Location
Chr.XII: 440467 - 441288 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182037.1 NP_013251.1 1 - 1 746 99

View Full Product Details