Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IDP2
Entrez Gene ID
850871
Gene Name
Gene Aliases
YLR174W
Chromosome Location
Chr.XII: 504591 - 505829 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182061.1 NP_013275.1 1 - 1 1152 88

View Full Product Details