Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEX13
Entrez Gene ID
850888
Gene Name
Gene Aliases
PAS20, YLR191W
Chromosome Location
Chr.XII: 537272 - 538432 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182078.1 NP_013292.1 1 - 1 1045 92

View Full Product Details