Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPR6
Entrez Gene ID
850914
Gene Name
Gene Aliases
CYP40, YLR216C
Chromosome Location
Chr.XII: 572096 - 573211 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182103.1 NP_013317.1 1 - 1 650 117

View Full Product Details