Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEC3
Entrez Gene ID
850995
Gene Name
Gene Aliases
PIP3, PSO9, YLR288C
Chromosome Location
Chr.XII: 713480 - 714904 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182176.1 NP_013391.1 1 - 1 1350 84

View Full Product Details