Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEX30
Entrez Gene ID
851034
Gene Name
Gene Aliases
YLR324W
Chromosome Location
Chr.XII: 779215 - 780786 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182213.1 NP_013428.1 1 - 1 1453 81

View Full Product Details