Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMA10
Entrez Gene ID
851037
Gene Name
Gene Aliases
SFL2, YLR327C
Chromosome Location
Chr.XII: 783127 - 783387 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182216.1 NP_013431.1 1 - 1 44 77

View Full Product Details