Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BUD8
Entrez Gene ID
851067
Gene Name
Gene Aliases
YLR353W
Chromosome Location
Chr.XII: 834352 - 836163 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182242.1 NP_013457.1 1 - 1 1752 64

View Full Product Details