Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STP3
Entrez Gene ID
851089
Gene Name
Gene Aliases
YLR375W
Chromosome Location
Chr.XII: 871697 - 872728 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182264.3 NP_013479.3 1 - 1 855 87

View Full Product Details