Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS33
Entrez Gene ID
851112
Gene Name
Gene Aliases
CLS14, MET27, PEP14, SLP1, VAM5, VPL25, VPT33, YLR396C
Chromosome Location
Chr.XII: 910235 - 912310 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182284.3 NP_013500.3 1 - 1 1850 68

View Full Product Details