Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL31B
Entrez Gene ID
851122
Gene Name
Gene Aliases
YLR406C
Chromosome Location
Chr.XII: 931065 - 931755 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182294.3 NP_013510.3 1 - 2 59 84

View Full Product Details