Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERV25
Entrez Gene ID
854997
Gene Name
Gene Aliases
YML012W
Chromosome Location
Chr.XIII: 246116 - 246751 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182369.1 NP_013701.1 1 - 1 451 100

View Full Product Details