Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRM9
Entrez Gene ID
854994
Gene Name
Gene Aliases
KTI1, YML014W
Chromosome Location
Chr.XIII: 243225 - 244064 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182372.1 NP_013698.1 1 - 1 755 99

View Full Product Details