Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRP39
Entrez Gene ID
854960
Gene Name
Gene Aliases
YML046W
Chromosome Location
Chr.XIII: 181474 - 183363 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182403.1 NP_013667.1 1 - 1 1745 93

View Full Product Details