Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RSE1
Entrez Gene ID
854956
Gene Name
Gene Aliases
YML049C
Chromosome Location
Chr.XIII: 174220 - 178305 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182407.1 NP_013663.1 1 - 1 3850 89

View Full Product Details