Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CMP2
Entrez Gene ID
854946
Gene Name
Gene Aliases
CNA2, YML057W
Chromosome Location
Chr.XIII: 160180 - 161994 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182415.1 NP_013655.1 1 - 1 1153 72

View Full Product Details