Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAP42
Entrez Gene ID
855043
Gene Name
Gene Aliases
YMR028W
Chromosome Location
Chr.XIII: 327482 - 328582 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182524.1 NP_013741.1 1 - 1 1052 83

View Full Product Details