Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ISF1
Entrez Gene ID
855105
Gene Name
Gene Aliases
MBR3, YMR081C
Chromosome Location
Chr.XIII: 430079 - 431095 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182580.1 NP_013798.1 1 - 1 946 99

View Full Product Details