Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ECM5
Entrez Gene ID
855214
Gene Name
Gene Aliases
YMR176W
Chromosome Location
Chr.XIII: 611740 - 615975 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182682.1 NP_013901.1 1 - 1 3945 98

View Full Product Details