Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ROT1
Entrez Gene ID
855240
Gene Name
Gene Aliases
YMR200W
Chromosome Location
Chr.XIII: 664752 - 665522 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182707.1 NP_013927.1 1 - 1 647 94

View Full Product Details