Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAS3
Entrez Gene ID
855255
Gene Name
Gene Aliases
YMR215W
Chromosome Location
Chr.XIII: 696796 - 698370 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182722.1 NP_013942.1 1 - 1 1447 111

View Full Product Details