Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADH2
Entrez Gene ID
855349
Gene Name
Gene Aliases
ADR2, YMR303C
Chromosome Location
Chr.XIII: 873291 - 874337 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182812.1 NP_014032.1 1 - 1 948 89

View Full Product Details