Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EOS1
Entrez Gene ID
855644
Gene Name
Gene Aliases
YNL080C
Chromosome Location
Chr.XIV: 476932 - 478032 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182918.1 NP_014319.1 1 - 1 1050 70

View Full Product Details